missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrobenzoic acid, 98%

Catalog No. p-7021976
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1453606 5 g
AAA1453614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1453606 Supplier Thermo Scientific Chemicals Supplier No. A1453606

CAS: 610-30-0 | C7H4N2O6 | 212.117 g/mol

2,4-Dinitrobenzoic acid was used in the spectrophotometric determination of diazepam in pure samples and in its pharmaceutical preparations. It was employed as chromophore probe for analysis of perfluorinated carboxylic acids in water by capillary zone electrophoresis. It can be used to produce 2,4-dinitro-benzoyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dinitrobenzoic acid was used in the spectrophotometric determination of diazepam in pure samples and in its pharmaceutical preparations. It was employed as chromophore probe for analysis of perfluorinated carboxylic acids in water by capillary zone electrophoresis. It can be used to produce 2,4-dinitro-benzoyl chloride.

Solubility
Soluble in dioxane 50 mg/ml.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 610-30-0
Molecular Formula C7H4N2O6
Molecular Weight (g/mol) 212.117
MDL Number MFCD00017006
InChI Key ZIIGSRYPZWDGBT-UHFFFAOYSA-N
Synonym 2,4-dinitrobenzoic acid, benzoic acid, 2,4-dinitro, ccris 1766, acmc-20aoc2, 2.4-dinitrobenzoic acid, benzoicacid, 2,4-dinitro, 2,4-dinitrobenzoic acidanion, 4-09-00-01239 beilstein handbook reference, aronis26960, 1-carboxy-2,4-dinitrobenzene
PubChem CID 11880
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)O

Specifications

Melting Point 178°C to 184°C
Quantity 5 g
Beilstein 658650
Solubility Information Soluble in dioxane 50mg/ml.
Formula Weight 212.12
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-219-5
RTECSNumber DG9140500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only