missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrobenzenesulfonic acid sodium salt, 97%

Catalog No. AAA1849314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1849314 25 g
AAA1849306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1849314 Supplier Thermo Scientific Chemicals Supplier No. A1849314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 885-62-1 | C6H3N2NaO7S | 270.15 g/mol

2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 885-62-1
Molecular Formula C6H3N2NaO7S
Molecular Weight (g/mol) 270.15
MDL Number MFCD00007471,MFCD10567393
InChI Key GSBYVRKLPCSLNV-UHFFFAOYSA-M
Synonym sodium 2,4-dinitrobenzenesulfonate hydrate, acmc-209quh, sodium dnbs hydrate, 2,4-dinitrobenzenesulfonic acid sodium salt hydrate
PubChem CID 70700115
IUPAC Name sodium;2,4-dinitrobenzenesulfonate;hydrate
SMILES [Na+].[O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O

Specifications

Quantity 25 g
Beilstein 6676807
Solubility Information Soluble in water and methanol.
Formula Weight 270.15
Percent Purity 97%
Chemical Name or Material 2,4-Dinitrobenzenesulfonic acid sodium salt
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-943-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only