missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitroaniline 98.0+%, TCI America™

Catalog No. p-7126448
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D081425G 25 g
D0814500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D081425G Supplier TCI America Supplier No. D081425G
Only null left

Chemical Identifiers

CAS 97-02-9
Molecular Formula C6H5N3O4
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007151
InChI Key LXQOQPGNCGEELI-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dinitro, 1-amino-2,4-dinitrobenzene, 2,4-dinitroanilin, 2,4-dinitrobenzenamine, 2,4-dinitroanilina, 2,4-dinitraniline, 2,4-dinitrophenylamine, aniline, 2,4-dinitro, 2,4-dinitroaminobenzene, 2,4-nitroaniline
PubChem CID 7321
ChEBI CHEBI:34242
IUPAC Name 2,4-dinitroaniline
SMILES NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 180°C
Color Yellow
Quantity 25 g
UN Number 1596
Formula Weight 183.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitroaniline
EINECSNumber (3)-0403
RTECSNumber BX9100000
TSCA Yes