Learn More
2,4-Dimethylstyrene, 97%, stabilized
CAS: 2234-20-0 | C10H12 | 132.21 g/mol
$141.88 - $141.88
Chemical Identifiers
| CAS | 2234-20-0 |
|---|---|
| Molecular Formula | C10H12 |
| Molecular Weight (g/mol) | 132.21 |
| MDL Number | MFCD00014937 |
| InChI Key | OEVVKKAVYQFQNV-UHFFFAOYSA-N |
| Synonym | 2,4-dimethylstyrene, 4-vinyl-m-xylene, styrene, 2,4-dimethyl, 1,3-dimethyl-4-vinylbenzene, 1-vinyl-2,4-dimethylbenzene, 1,3-dimethyl-4-ethenylbenzene, benzene, 1-ethenyl-2,4-dimethyl, unii-543es1o25e, styrene,4-dimethyl, acmc-1cb1s |
| PubChem CID | 16694 |
| IUPAC Name | 1-ethenyl-2,4-dimethylbenzene |
| SMILES | CC1=CC(=C(C=C1)C=C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC271150050
|
Thermo Scientific Chemicals
271150050 |
5 g | Glass bottle |
Each for $141.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2234-20-0 | |
| 132.21 | |
| OEVVKKAVYQFQNV-UHFFFAOYSA-N | |
| 16694 | |
| CC1=CC(=C(C=C1)C=C)C |
| C10H12 | |
| MFCD00014937 | |
| 2,4-dimethylstyrene, 4-vinyl-m-xylene, styrene, 2,4-dimethyl, 1,3-dimethyl-4-vinylbenzene, 1-vinyl-2,4-dimethylbenzene, 1,3-dimethyl-4-ethenylbenzene, benzene, 1-ethenyl-2,4-dimethyl, unii-543es1o25e, styrene,4-dimethyl, acmc-1cb1s | |
| 1-ethenyl-2,4-dimethylbenzene |
Specifications
| 2234-20-0 | |
| 100.0 | |
| 0.9060g/mL | |
| 60°C | |
| 96% min. (GC) | |
| C10H12 | |
| (CH3)2C6H3CH=CH2 | |
| 5 g | |
| 2,4-dimethylstyrene, 4-vinyl-m-xylene, styrene, 2,4-dimethyl, 1,3-dimethyl-4-vinylbenzene, 1-vinyl-2,4-dimethylbenzene, 1,3-dimethyl-4-ethenylbenzene, benzene, 1-ethenyl-2,4-dimethyl, unii-543es1o25e, styrene,4-dimethyl, acmc-1cb1s | |
| CC1=CC(=C(C=C1)C=C)C | |
| 132.21 | |
| 16694 | |
| 97% | |
| 2, 4-Dimethylstyrene, 97% |
| 96.0 | |
| -64.0°C | |
| 73.0°C to 74.0°C (10.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.5410 to 1.5440 | |
| MFCD00014937 | |
| 0.906 | |
| OEVVKKAVYQFQNV-UHFFFAOYSA-N | |
| 1-ethenyl-2,4-dimethylbenzene | |
| 0.05 % (ca.) t-butylcatechol | |
| 132.21 | |
| Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr
GHS Signal Word: Warning
EINECSNumber : 218-781-3
RTECSNumber : WL4450000
TSCA : TSCA
RUO – Research Use Only