missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethylpyrrole 97.0+%, TCI America™
$71.23 - $762.36
Chemical Identifiers
| CAS | 625-82-1 |
|---|---|
| Molecular Formula | C6H9N |
| Molecular Weight (g/mol) | 95.15 |
| MDL Number | MFCD00192088 |
| InChI Key | MFFMQGGZCLEMCI-UHFFFAOYSA-N |
| Synonym | 2,4-dimethylpyrrole, 1h-pyrrole, 2,4-dimethyl, pyrrole, 2,4-dimethyl, unii-ynq49m599x, pubchem24003, 2,4-dimethyl-1h-pyrrol, acmc-209n5e, ksc493o6j, # |
| PubChem CID | 39539 |
| IUPAC Name | 2,4-dimethyl-1H-pyrrole |
| SMILES | CC1=CC(C)=CN1 |
Chemical Identifiers
| 625-82-1 | |
| 95.15 | |
| MFFMQGGZCLEMCI-UHFFFAOYSA-N | |
| 39539 | |
| CC1=CC(C)=CN1 |
| C6H9N | |
| MFCD00192088 | |
| 2,4-dimethylpyrrole, 1h-pyrrole, 2,4-dimethyl, pyrrole, 2,4-dimethyl, unii-ynq49m599x, pubchem24003, 2,4-dimethyl-1h-pyrrol, acmc-209n5e, ksc493o6j, # | |
| 2,4-dimethyl-1H-pyrrole |
Specifications
| 625-82-1 | |
| MFCD00192088 | |
| 2,4-dimethylpyrrole, 1h-pyrrole, 2,4-dimethyl, pyrrole, 2,4-dimethyl, unii-ynq49m599x, pubchem24003, 2,4-dimethyl-1h-pyrrol, acmc-209n5e, ksc493o6j, # | |
| CC1=CC(C)=CN1 | |
| 95.15 | |
| 95.15 | |
| Liquid |
| C6H9N | |
| 1 g | |
| MFFMQGGZCLEMCI-UHFFFAOYSA-N | |
| 2,4-dimethyl-1H-pyrrole | |
| 39539 | |
| ≥97.0% (GC) | |
| 2,4-Dimethylpyrrole |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator