accessibility menu, dialog, popup

UserName

CH3NHH3C

CAS RN 625-82-1

IUPAC Name: 2,4-dimethyl-1H-pyrrole
Synonyms: 1H-pyrrole, 2,4-dimethyl-
Molecular Weight (g/mol): 95.15
Molecular Formula: C6H9N
InChi Key: MFFMQGGZCLEMCI-UHFFFAOYSA-N
SMILES: CC1=CC(C)=CN1
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)