missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylpyrrole 97.0+%, TCI America™

Catalog No. D28481G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28481G 1 g
D28485G 5 g
D284825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D28481G Supplier TCI America Supplier No. D28481G

Chemical Identifiers

CAS 625-82-1
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.15
MDL Number MFCD00192088
InChI Key MFFMQGGZCLEMCI-UHFFFAOYSA-N
Synonym 2,4-dimethylpyrrole, 1h-pyrrole, 2,4-dimethyl, pyrrole, 2,4-dimethyl, unii-ynq49m599x, pubchem24003, 2,4-dimethyl-1h-pyrrol, acmc-209n5e, ksc493o6j, #
PubChem CID 39539
IUPAC Name 2,4-dimethyl-1H-pyrrole
SMILES CC1=CC(C)=CN1

Specifications

Quantity 1 g
Formula Weight 95.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylpyrrole
TSCA No
Recommended Storage Refrigerator