missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylpyrrole, 97%

Catalog No. AC319650250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC319650250 25 g
AC319650010 1 g
AC319650050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC319650250 Supplier Thermo Scientific Chemicals Supplier No. 319650250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 625-82-1 | C6H9N | 95.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 625-82-1
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.15
MDL Number MFCD00192088
InChI Key MFFMQGGZCLEMCI-UHFFFAOYSA-N
Synonym 2,4-dimethylpyrrole, 1h-pyrrole, 2,4-dimethyl, pyrrole, 2,4-dimethyl, unii-ynq49m599x, pubchem24003, 2,4-dimethyl-1h-pyrrol, acmc-209n5e, ksc493o6j, #
PubChem CID 39539
IUPAC Name 2,4-dimethyl-1H-pyrrole
SMILES CC1=CC(C)=CN1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown-Maroon to Orange-Red or Violet
Density 0.9240g/mL
Boiling Point 165.0°C to 167.0°C
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Refractive Index 1.4950 to 1.4970
Quantity 25 g
Specific Gravity 0.924
Formula Weight 95.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dimethylpyrrole, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-912-2
TSCA TSCA
Recommended Storage May darken on storage

RUO – Research Use Only