missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D35141G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35141G Supplier TCI America Supplier No. D35141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55499-44-0 | C8H11BO2 | 149.984 g/mol

Specifications

Chemical Name or Material 2,4-Dimethylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 55499-44-0
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C8H11BO2
MDL Number MFCD02683101
Synonym 2,4-dimethylphenyl boronic acid, 2,4-dimethylbenzeneboronic acid, 2,4-dimethyl phenyl boronic acid, 2,4-dimethylphenylboronicacid, 4-dimethylphenylboronic acid, boronic acid, 2,4-dimethylphenyl, 4-borono-m-xylene, pubchem9565, m-xylene-4-boronic acid, acmc-1ay9d
InChI Key TYONHSPZXLFWKI-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)C)C)(O)O
IUPAC Name (2,4-dimethylphenyl)boronic acid
Molecular Weight (g/mol) 149.984
PubChem CID 4198739
Formula Weight 149.98
Percent Purity 97.0 to 116.0%
Grade Not Available
Show More Show Less