missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyloxazole 98.0+%, TCI America™

Catalog No. D37665G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37665G 5 g
D37661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37665G Supplier TCI America Supplier No. D37665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7208-05-1
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.12
MDL Number MFCD00234124
InChI Key PSOZJOZKEVZLKZ-UHFFFAOYSA-N
PubChem CID 138961
IUPAC Name 2,4-dimethyl-1,3-oxazole
SMILES CC1=COC(=N1)C

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
UN Number 1993
Formula Weight 97.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethyloxazole