accessibility menu, dialog, popup

UserName

CH3OH3CN

CAS RN 7208-05-1

IUPAC Name: 2,4-dimethyl-1,3-oxazole
Synonyms: oxazole, 2,4-dimethyl-
Molecular Weight (g/mol): 97.12
Molecular Formula: C5H7NO
InChi Key: PSOZJOZKEVZLKZ-UHFFFAOYSA-N
SMILES: CC1=NC(C)=CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results