missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzophenone 96.0+%, TCI America™

Catalog No. D345825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D345825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D345825G Supplier TCI America Supplier No. D345825G

Chemical Identifiers

CAS 1140-14-3
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00008537
InChI Key UYSQHMXRROFKRN-UHFFFAOYSA-N
PubChem CID 70831
IUPAC Name (2,4-dimethylphenyl)-phenylmethanone
SMILES CC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)C

Specifications

Color Yellow
Boiling Point 189°C
Quantity 25 g
Formula Weight 210.28
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylbenzophenone