missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzoic acid, 97%

Catalog No. p-7080849
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1058909 10 g
AAA1058918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1058909 Supplier Thermo Scientific Chemicals Supplier No. A1058909
Only null left

CAS: 611-01-8 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-01-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002480
InChI Key BKYWPNROPGQIFZ-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dimethyl, 4-carboxy-1,3-dimethylbenzene, 2,4-dimethyl benzoic acid, m-xylene-4-carboxylic acid, 2,4-dimethyl-benzoic acid, 2,4-dimethylbenzoicacid, m-xylylic acid, pubchem15441, 2,4 dimethylbenzoic acid, benzoic acid,4-dimethyl
PubChem CID 11897
ChEBI CHEBI:64811
IUPAC Name 2,4-dimethylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(C)=C1

Specifications

Melting Point 124°C to 126°C
Quantity 10 g
Odor Odorless
Beilstein 971589
Boiling Point 267°C (727 mmHg)
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2,4-Dimethylbenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-246-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only