missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzeneboronic acid, 97%

Catalog No. AAB2307603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2307603 1 g
AAB2307606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2307603 Supplier Thermo Scientific Chemicals Supplier No. B2307603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 55499-44-0 | C8H11BO2 | 149.984 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55499-44-0
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.984
MDL Number MFCD02683101
InChI Key TYONHSPZXLFWKI-UHFFFAOYSA-N
Synonym 2,4-dimethylphenyl boronic acid, 2,4-dimethylbenzeneboronic acid, 2,4-dimethyl phenyl boronic acid, 2,4-dimethylphenylboronicacid, 4-dimethylphenylboronic acid, boronic acid, 2,4-dimethylphenyl, 4-borono-m-xylene, pubchem9565, m-xylene-4-boronic acid, acmc-1ay9d
PubChem CID 4198739
IUPAC Name (2,4-dimethylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)C)C)(O)O

Specifications

Melting Point 204°C to 208°C
Quantity 1 g
Formula Weight 149.99
Percent Purity 97%
Chemical Name or Material 2,4-Dimethylbenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only