missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethylbenzaldehyde 97.0+%, TCI America™
$89.48 - $602.10
Chemical Identifiers
| CAS | 15764-16-6 |
|---|---|
| Molecular Formula | C9H10O |
| Molecular Weight (g/mol) | 134.178 |
| MDL Number | MFCD00003340 |
| InChI Key | GISVICWQYMUPJF-UHFFFAOYSA-N |
| Synonym | benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde |
| PubChem CID | 61814 |
| IUPAC Name | 2,4-dimethylbenzaldehyde |
| SMILES | CC1=CC(=C(C=C1)C=O)C |
Chemical Identifiers
| 15764-16-6 | |
| 134.178 | |
| GISVICWQYMUPJF-UHFFFAOYSA-N | |
| 61814 | |
| CC1=CC(=C(C=C1)C=O)C |
| C9H10O | |
| MFCD00003340 | |
| benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde | |
| 2,4-dimethylbenzaldehyde |
Specifications
| 15764-16-6 | |
| 98°C | |
| MFCD00003340 | |
| benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde | |
| CC1=CC(=C(C=C1)C=O)C | |
| 134.178 | |
| 134.18 | |
| Liquid |
| Yellow | |
| C9H10O | |
| 25 g | |
| GISVICWQYMUPJF-UHFFFAOYSA-N | |
| 2,4-dimethylbenzaldehyde | |
| 61814 | |
| ≥97.0% (GC) | |
| 2,4-Dimethylbenzaldehyde |
Safety and Handling
EINECSNumber : (3)-3763
TSCA : No