missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzaldehyde 97.0+%, TCI America™

Catalog No. D239225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D239225G 25 g
D2392500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D239225G Supplier TCI America Supplier No. D239225G

Chemical Identifiers

CAS 15764-16-6
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00003340
InChI Key GISVICWQYMUPJF-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde
PubChem CID 61814
IUPAC Name 2,4-dimethylbenzaldehyde
SMILES CC1=CC(=C(C=C1)C=O)C

Specifications

Color Yellow
Boiling Point 98°C
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylbenzaldehyde
EINECSNumber (3)-3763
TSCA No