missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzaldehyde, 99%

Catalog No. AC165790050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC165790050 5 g Glass bottle
AC165790250 25 g Glass bottle
AC165791000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC165790050 Supplier Thermo Scientific Chemicals Supplier No. 165790050

CAS: 15764-16-6 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15764-16-6
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003340
InChI Key GISVICWQYMUPJF-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde
PubChem CID 61814
IUPAC Name 2,4-dimethylbenzaldehyde
SMILES CC1=CC(=C(C=C1)C=O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -9°C
Color Colorless
Density 0.9600g/mL
Boiling Point 102.4°C to 103°C (14.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5482 to 1.5502
Linear Formula (CH3)2C6H3CHO
Quantity 5 g
Beilstein 07, IV, 706
Specific Gravity 0.96
Formula Weight 134.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethylbenzaldehyde
Show More Show Less
EINECSNumber 239-856-7

RUO – Research Use Only