missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethylbenzaldehyde, 90+%
CAS: 15764-16-6 | C9H10O | 134.178 g/mol
$87.22 - $296.58
Chemical Identifiers
| CAS | 15764-16-6 |
|---|---|
| Molecular Formula | C9H10O |
| Molecular Weight (g/mol) | 134.178 |
| MDL Number | MFCD00003340 |
| InChI Key | GISVICWQYMUPJF-UHFFFAOYSA-N |
| Synonym | benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde |
| PubChem CID | 61814 |
| IUPAC Name | 2,4-dimethylbenzaldehyde |
| SMILES | CC1=CC(=C(C=C1)C=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1465506
|
Thermo Scientific Chemicals
A1465506 |
5 g |
Each for $87.22
|
|
|||||
|
AAA1465514
|
Thermo Scientific Chemicals
A1465514 |
25 g |
Each for $296.58
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15764-16-6 | |
| 134.178 | |
| GISVICWQYMUPJF-UHFFFAOYSA-N | |
| 61814 | |
| CC1=CC(=C(C=C1)C=O)C |
| C9H10O | |
| MFCD00003340 | |
| benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde | |
| 2,4-dimethylbenzaldehyde |
Specifications
| 15764-16-6 | |
| 1.02 | |
| 88°C (190°F) | |
| C9H10O | |
| MFCD00003340 | |
| Air Sensitive | |
| GISVICWQYMUPJF-UHFFFAOYSA-N | |
| 2,4-dimethylbenzaldehyde | |
| 61814 | |
| ≥90% |
| -9°C | |
| 215°C to 216°C | |
| Almond-like | |
| 1.549 | |
| 5 g | |
| benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde | |
| CC1=CC(=C(C=C1)C=O)C | |
| 134.178 | |
| 134.18 | |
| 2,4-Dimethylbenzaldehyde |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 239-856-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only