missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzaldehyde, 90+%

Catalog No. AAA1465514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1465514 25 g
AAA1465506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1465514 Supplier Thermo Scientific Chemicals Supplier No. A1465514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15764-16-6 | C9H10O | 134.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15764-16-6
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00003340
InChI Key GISVICWQYMUPJF-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde
PubChem CID 61814
IUPAC Name 2,4-dimethylbenzaldehyde
SMILES CC1=CC(=C(C=C1)C=O)C

Specifications

Melting Point -9°C
Density 1.02
Boiling Point 215°C to 216°C
Flash Point 88°C (190°F)
Odor Almond-like
Refractive Index 1.549
Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 134.18
Percent Purity ≥90%
Chemical Name or Material 2,4-Dimethylbenzaldehyde
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 239-856-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only