missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylaniline 98.0+%, TCI America™

Catalog No. p-7126358
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D066725ML 25 mL
D0667500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D066725ML Supplier TCI America Supplier No. D066725ML

Chemical Identifiers

CAS 95-68-1
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007738
InChI Key CZZZABOKJQXEBO-UHFFFAOYSA-N
Synonym 2,4-xylidine, 2,4-dimethyl aniline, m-xylidine, m-4-xylidine, 2,4-dimethylphenylamine, benzenamine, 2,4-dimethyl, 2-methyl-p-toluidine, 4-methyl-o-toluidine, 1-amino-2,4-dimethylbenzene, 4-amino-1,3-xylene
PubChem CID 7250
ChEBI CHEBI:27840
IUPAC Name 2,4-dimethylaniline
SMILES CC1=CC(=C(C=C1)N)C

Specifications

Melting Point 16°C
Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1711
Boiling Point 214°C
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2,4-Dimethylaniline
EINECSNumber (3)-0129
RTECSNumber ZE8925000
TSCA Yes