missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylaniline-5-sulfonic Acid Sodium Salt Hydrate, TCI America™

Catalog No. X003025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X003025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X003025G Supplier TCI America Supplier No. X003025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64501-84-4
Molecular Formula C8H10NNaO3S
Molecular Weight (g/mol) 223.222
MDL Number MFCD00070550
InChI Key ZETYURDDFWHYDN-UHFFFAOYSA-M
Synonym m-Xylidine-5-sulfonic Acid Sodium Salt
PubChem CID 23691697
IUPAC Name sodium;5-amino-2,4-dimethylbenzenesulfonate
SMILES CC1=CC(=C(C=C1N)S(=O)(=O)[O-])C.[Na+]

Specifications

Quantity 25 g
Formula Weight 223.22
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethylaniline-5-sulfonic Acid Sodium Salt Hydrate