missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4'-Dimethylacetophenone 95.0+%, TCI America™
Supplier: TCI America D18795ML
Specifications
| 2′,4′-Dimethylacetophenone | |
| Yellow | |
| C10H12O | |
| 5 mL | |
| HSDSKVWQTONQBJ-UHFFFAOYSA-N | |
| 1-(2,4-dimethylphenyl)ethanone | |
| 6985 | |
| ≥95.0% (GC) |
| 89-74-7 | |
| 122°C | |
| MFCD00003571 | |
| 2',4'-dimethylacetophenone, 1-2,4-dimethylphenyl ethanone, 2,4-dimethylacetophenone, 4-acetyl-m-xylene, ethanone, 1-2,4-dimethylphenyl, acetyl-m-xylene, 1-2,4-dimethylphenyl ethan-1-one, acetophenone, 2',4'-dimethyl, unii-8k29me27ya, methyl 2,4-dimethylphenyl ketone | |
| CC1=CC(=C(C=C1)C(=O)C)C | |
| 148.205 | |
| 148.21 | |
| Liquid |
Chemical Identifiers
| 89-74-7 | |
| 148.205 | |
| HSDSKVWQTONQBJ-UHFFFAOYSA-N | |
| 6985 | |
| CC1=CC(=C(C=C1)C(=O)C)C |
| C10H12O | |
| MFCD00003571 | |
| 2',4'-dimethylacetophenone, 1-2,4-dimethylphenyl ethanone, 2,4-dimethylacetophenone, 4-acetyl-m-xylene, ethanone, 1-2,4-dimethylphenyl, acetyl-m-xylene, 1-2,4-dimethylphenyl ethan-1-one, acetophenone, 2',4'-dimethyl, unii-8k29me27ya, methyl 2,4-dimethylphenyl ketone | |
| 1-(2,4-dimethylphenyl)ethanone |
Safety and Handling
TSCA : Yes