missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethylacetophenone 95.0+%, TCI America™

Catalog No. p-7126988
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18795ML 5 mL
D187925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18795ML Supplier TCI America Supplier No. D18795ML
Only null left

Chemical Identifiers

CAS 89-74-7
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00003571
InChI Key HSDSKVWQTONQBJ-UHFFFAOYSA-N
Synonym 2',4'-dimethylacetophenone, 1-2,4-dimethylphenyl ethanone, 2,4-dimethylacetophenone, 4-acetyl-m-xylene, ethanone, 1-2,4-dimethylphenyl, acetyl-m-xylene, 1-2,4-dimethylphenyl ethan-1-one, acetophenone, 2',4'-dimethyl, unii-8k29me27ya, methyl 2,4-dimethylphenyl ketone
PubChem CID 6985
IUPAC Name 1-(2,4-dimethylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)C(=O)C)C

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 mL
Formula Weight 148.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,4′-Dimethylacetophenone