Learn More
2',4'-Dimethylacetophenone, 95%
CAS: 89-74-7 | C10H12O | 148.205 g/mol
Supplier: Thermo Scientific Chemicals A1299209
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2',4'-Dimethylacetophenone | |
| 0.999 | |
| 100°C (212°F) | |
| C10H12O | |
| MFCD00003571 | |
| 956772 | |
| HSDSKVWQTONQBJ-UHFFFAOYSA-N | |
| 1-(2,4-dimethylphenyl)ethanone | |
| 6985 | |
| 97% |
| 89-74-7 | |
| 227°C to 228°C | |
| Characteristic | |
| 1.535 | |
| 10 g | |
| 2',4'-dimethylacetophenone, 1-2,4-dimethylphenyl ethanone, 2,4-dimethylacetophenone, 4-acetyl-m-xylene, ethanone, 1-2,4-dimethylphenyl, acetyl-m-xylene, 1-2,4-dimethylphenyl ethan-1-one, acetophenone, 2',4'-dimethyl, unii-8k29me27ya, methyl 2,4-dimethylphenyl ketone | |
| CC1=CC(=C(C=C1)C(=O)C)C | |
| 148.205 | |
| 148.21 |
Chemical Identifiers
| 89-74-7 | |
| 148.205 | |
| HSDSKVWQTONQBJ-UHFFFAOYSA-N | |
| 6985 | |
| CC1=CC(=C(C=C1)C(=O)C)C |
| C10H12O | |
| MFCD00003571 | |
| 2',4'-dimethylacetophenone, 1-2,4-dimethylphenyl ethanone, 2,4-dimethylacetophenone, 4-acetyl-m-xylene, ethanone, 1-2,4-dimethylphenyl, acetyl-m-xylene, 1-2,4-dimethylphenyl ethan-1-one, acetophenone, 2',4'-dimethyl, unii-8k29me27ya, methyl 2,4-dimethylphenyl ketone | |
| 1-(2,4-dimethylphenyl)ethanone |
Safety and Handling
P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
H302+H332-H319
EINECSNumber : 201-935-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only