missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethylacetophenone, 95%

Catalog No. p-7020885
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1299209 10 g
AAA1299218 50 g
AAA1299230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1299209 Supplier Thermo Scientific Chemicals Supplier No. A1299209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89-74-7 | C10H12O | 148.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-74-7
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00003571
InChI Key HSDSKVWQTONQBJ-UHFFFAOYSA-N
Synonym 2',4'-dimethylacetophenone, 1-2,4-dimethylphenyl ethanone, 2,4-dimethylacetophenone, 4-acetyl-m-xylene, ethanone, 1-2,4-dimethylphenyl, acetyl-m-xylene, 1-2,4-dimethylphenyl ethan-1-one, acetophenone, 2',4'-dimethyl, unii-8k29me27ya, methyl 2,4-dimethylphenyl ketone
PubChem CID 6985
IUPAC Name 1-(2,4-dimethylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)C(=O)C)C

Specifications

Density 0.999
Boiling Point 227°C to 228°C
Flash Point 100°C (212°F)
Odor Characteristic
Refractive Index 1.535
Quantity 10 g
Beilstein 956772
Formula Weight 148.21
Percent Purity 97%
Chemical Name or Material 2',4'-Dimethylacetophenone
Hazard Category H302+H332-H319
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 201-935-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only