missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethyl-6-nitrophenol 98.0+%, TCI America™
$130.48 - $439.92
Chemical Identifiers
| CAS | 14452-34-7 |
|---|---|
| Molecular Formula | C8H9NO3 |
| Molecular Weight (g/mol) | 167.16 |
| MDL Number | MFCD00191633 |
| InChI Key | KJRCHILWKQLEBC-UHFFFAOYSA-N |
| Synonym | 6-nitro-2,4-xylenol, phenol, 2,4-dimethyl-6-nitro, acmc-1buww, intermediates-zcf02108, 2-nitro-4,6-dimethylphenol, ksc524c1l, phenol,2,4-dimethyl-6-nitro, 4,6-dimethyl-2-nitrophenol, 2,4-dimethyl-6-nitrophenol 2-nitro-4,6-dimethylphenol |
| PubChem CID | 84450 |
| IUPAC Name | 2,4-dimethyl-6-nitrophenol |
| SMILES | CC1=CC(C)=C(O)C(=C1)[N+]([O-])=O |
Chemical Identifiers
| 14452-34-7 | |
| 167.16 | |
| KJRCHILWKQLEBC-UHFFFAOYSA-N | |
| 84450 | |
| CC1=CC(C)=C(O)C(=C1)[N+]([O-])=O |
| C8H9NO3 | |
| MFCD00191633 | |
| 6-nitro-2,4-xylenol, phenol, 2,4-dimethyl-6-nitro, acmc-1buww, intermediates-zcf02108, 2-nitro-4,6-dimethylphenol, ksc524c1l, phenol,2,4-dimethyl-6-nitro, 4,6-dimethyl-2-nitrophenol, 2,4-dimethyl-6-nitrophenol 2-nitro-4,6-dimethylphenol | |
| 2,4-dimethyl-6-nitrophenol |
Specifications
| 71°C | |
| 5 g | |
| C8H9NO3 | |
| 6-nitro-2,4-xylenol, phenol, 2,4-dimethyl-6-nitro, acmc-1buww, intermediates-zcf02108, 2-nitro-4,6-dimethylphenol, ksc524c1l, phenol,2,4-dimethyl-6-nitro, 4,6-dimethyl-2-nitrophenol, 2,4-dimethyl-6-nitrophenol 2-nitro-4,6-dimethylphenol | |
| CC1=CC(C)=C(O)C(=C1)[N+]([O-])=O | |
| 167.16 | |
| 167.16 | |
| 2,4-Dimethyl-6-nitrophenol |
| 14452-34-7 | |
| Crystal-Powder at 20°C | |
| MFCD00191633 | |
| KJRCHILWKQLEBC-UHFFFAOYSA-N | |
| 2,4-dimethyl-6-nitrophenol | |
| 84450 | |
| ≥98.0% (GC,T) |
Safety and Handling
TSCA : Yes