missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-6-nitrophenol, 98%

Catalog No. AAB2520522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2520522 100 g
AAB2520506 5 g
AAB2520514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2520522 Supplier Thermo Scientific Chemicals Supplier No. B2520522
Only null left

CAS: 14452-34-7 | C8H9NO3 | 167.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14452-34-7
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00191633
InChI Key KJRCHILWKQLEBC-UHFFFAOYSA-N
PubChem CID 84450
IUPAC Name 2,4-dimethyl-6-nitrophenol
SMILES CC1=CC(C)=C(O)C(=C1)[N+]([O-])=O

Specifications

Melting Point 68°C to 71°C
Quantity 100 g
Synonym 6-nitro-2,4-xylenol, phenol, 2,4-dimethyl-6-nitro, acmc-1buww, intermediates-zcf02108, 2-nitro-4,6-dimethylphenol, ksc524c1l, phenol,2,4-dimethyl-6-nitro, 4,6-dimethyl-2-nitrophenol, 2,4-dimethyl-6-nitrophenol 2-nitro-4,6-dimethylphenol
Formula Weight 167.17
Percent Purity 98%
Chemical Name or Material 2,4-Dimethyl-6-nitrophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-436-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only