missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethyl-6-hydroxypyrimidine 98.0+%, TCI America™
$186.74 - $610.28
Chemical Identifiers
| CAS | 6622-92-0 |
|---|---|
| Molecular Formula | C6H8N2O |
| Molecular Weight (g/mol) | 124.143 |
| MDL Number | MFCD00006105 |
| InChI Key | UQFHLJKWYIJISA-UHFFFAOYSA-N |
| Synonym | 2,4-dimethyl-6-hydroxypyrimidine, 2,6-dimethylpyrimidin-4-ol, 4-hydroxy-2,6-dimethylpyrimidine, 6-hydroxy-2,4-dimethylpyrimidine, 2,6-dimethyl-4-pyrimidinol, 4 1h-pyrimidinone, 2,6-dimethyl, 2,6-dimethyl-4-hydroxypyrimidine, 2,6-dimethylpyrimidin-4 3h-one, 2,4-dimethyl-6-oxypyrimidine, t6vm dnj c1 e1 |
| PubChem CID | 81098 |
| IUPAC Name | 2,6-dimethyl-1H-pyrimidin-4-one |
| SMILES | CC1=CC(=O)N=C(N1)C |
Chemical Identifiers
| 6622-92-0 | |
| 124.143 | |
| UQFHLJKWYIJISA-UHFFFAOYSA-N | |
| 81098 | |
| CC1=CC(=O)N=C(N1)C |
| C6H8N2O | |
| MFCD00006105 | |
| 2,4-dimethyl-6-hydroxypyrimidine, 2,6-dimethylpyrimidin-4-ol, 4-hydroxy-2,6-dimethylpyrimidine, 6-hydroxy-2,4-dimethylpyrimidine, 2,6-dimethyl-4-pyrimidinol, 4 1h-pyrimidinone, 2,6-dimethyl, 2,6-dimethyl-4-hydroxypyrimidine, 2,6-dimethylpyrimidin-4 3h-one, 2,4-dimethyl-6-oxypyrimidine, t6vm dnj c1 e1 | |
| 2,6-dimethyl-1H-pyrimidin-4-one |
Specifications
| 6622-92-0 | |
| White-Yellow | |
| MFCD00006105 | |
| 2,4-dimethyl-6-hydroxypyrimidine, 2,6-dimethylpyrimidin-4-ol, 4-hydroxy-2,6-dimethylpyrimidine, 6-hydroxy-2,4-dimethylpyrimidine, 2,6-dimethyl-4-pyrimidinol, 4 1h-pyrimidinone, 2,6-dimethyl, 2,6-dimethyl-4-hydroxypyrimidine, 2,6-dimethylpyrimidin-4 3h-one, 2,4-dimethyl-6-oxypyrimidine, t6vm dnj c1 e1 | |
| CC1=CC(=O)N=C(N1)C | |
| 124.143 | |
| 124.14 | |
| Crystalline Powder |
| 199°C | |
| C6H8N2O | |
| 5 g | |
| UQFHLJKWYIJISA-UHFFFAOYSA-N | |
| 2,6-dimethyl-1H-pyrimidin-4-one | |
| 81098 | |
| ≥98.0% (HPLC,T) | |
| 2,4-Dimethyl-6-hydroxypyrimidine |
Safety and Handling
TSCA : No