missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-pentanone 98.0+%, TCI America™

Catalog No. p-7126421
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
D077025ML 25 mL
D0770500ML 500 mL
2 options

Catalog No. D077025ML

Supplier: TCI America D077025ML

Only null left

Chemical Identifiers

CAS 565-80-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008918
InChI Key HXVNBWAKAOHACI-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
PubChem CID 11271
IUPAC Name 2,4-dimethylpentan-3-one
SMILES CC(C)C(=O)C(C)C

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 125°C
Quantity 25 mL
UN Number 1224
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethyl-3-pentanone
EINECSNumber (2)-2475
RTECSNumber SA8575500
TSCA Yes