missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-pentanone 98.0+%, TCI America™

Catalog No. D077025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D077025ML 25 mL
D0770500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D077025ML Supplier TCI America Supplier No. D077025ML

Chemical Identifiers

CAS 565-80-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008918
InChI Key HXVNBWAKAOHACI-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
PubChem CID 11271
IUPAC Name 2,4-dimethylpentan-3-one
SMILES CC(C)C(=O)C(C)C

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 125°C
Quantity 25 mL
UN Number 1224
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethyl-3-pentanone
EINECSNumber (2)-2475
RTECSNumber SA8575500
TSCA Yes