missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-furoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC65801DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC65801DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

15058-72-7
140.138
2,4-dimethyl-3-furoic acid, 2,4-dimethyl-3-furancarboxylic acid, 3-furancarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-furan-3-carboxylic acid, 2,4-dimethylfurane-3-carboxylic acid
2,4-dimethylfuran-3-carboxylic acid
C7H8O3
JFXXYKVSLHRBAA-UHFFFAOYSA-N
638456
CC1=COC(=C1C(=O)O)C

Specifications

2, 4-Dimethyl-3-furoic acid
97%
C7H8O3
2,4-dimethyl-3-furoic acid, 2,4-dimethyl-3-furancarboxylic acid, 3-furancarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-furan-3-carboxylic acid, 2,4-dimethylfurane-3-carboxylic acid
CC1=COC(=C1C(=O)O)C
140.138
140.14
15058-72-7
Amber Glass Bottle
1g
JFXXYKVSLHRBAA-UHFFFAOYSA-N
2,4-dimethylfuran-3-carboxylic acid
638456
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents