missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1-pentene, 99%

Catalog No. p-7024787
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2000288 2.5 g
AAB2000209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2000288 Supplier Thermo Scientific Chemicals Supplier No. B2000288
Only null left

CAS: 2213-32-3 | C7H14 | 98.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2213-32-3
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00039854
InChI Key LXQPBCHJNIOMQU-UHFFFAOYSA-N
Synonym 2,4-dimethyl-1-pentene, 1-pentene, 2,4-dimethyl, acmc-1ch0z, pentene, 2,4-dimethyl, ch3 2chch2c ch3 =ch2, 1-pentene, 2,4-dimethyl-8ci 9ci
PubChem CID 16657
IUPAC Name 2,4-dimethylpent-1-ene
SMILES CC(C)CC(=C)C

Specifications

Density 0.69
Boiling Point 81°C
Flash Point −16°C (−27°F)
Refractive Index 1.3995
Quantity 2.5 g
UN Number UN3295
Formula Weight 98.19
Percent Purity 99%
Chemical Name or Material 2,4-Dimethyl-1-pentene
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 218-665-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only