missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1-hexene 98.0+%, TCI America™

Catalog No. D12415ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12415ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12415ML Supplier TCI America Supplier No. D12415ML
Only null left

Chemical Identifiers

CAS 16746-87-5
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00048655
InChI Key PKVDGQHNRICJLA-UHFFFAOYNA-N
PubChem CID 519301
IUPAC Name 2,4-dimethylhex-1-ene
SMILES CCC(C)CC(C)=C

Specifications

Color Colorless
Boiling Point 111°C
Quantity 5 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethyl-1-hexene
EINECSNumber (2)-0024
TSCA No