accessibility menu, dialog, popup

UserName

CAS RN 16746-87-5

H3CCH3CH3CH2

CAS RN 16746-87-5

IUPAC Name: 2,4-dimethylhex-1-ene
Synonyms: 2,4-dimethylhex-1-ene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: PKVDGQHNRICJLA-UHFFFAOYNA-N
SMILES: CCC(C)CC(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results