missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethyl-1-heptene 98.0+%, TCI America™
Supplier: TCI America D12581ML
Specifications
| 2,4-Dimethyl-1-heptene | |
| Colorless | |
| C9H18 | |
| 1 mL | |
| CZGAOHSMVSIJJZ-UHFFFAOYNA-N | |
| 2,4-dimethylhept-1-ene | |
| 123385 | |
| ≥98.0% (GC) |
| 19549-87-2 | |
| 134°C | |
| MFCD00059250 | |
| 3295 | |
| CCCC(C)CC(C)=C | |
| 126.24 | |
| 126.24 | |
| Liquid |
Chemical Identifiers
| 19549-87-2 | |
| 126.24 | |
| CZGAOHSMVSIJJZ-UHFFFAOYNA-N | |
| 2,4-dimethylhept-1-ene |
| C9H18 | |
| MFCD00059250 | |
| 123385 | |
| CCCC(C)CC(C)=C |
Safety and Handling
EINECSNumber : (2)-0025
TSCA : No