accessibility menu, dialog, popup

UserName

H3CCH3CH3CH2

CAS RN 19549-87-2

IUPAC Name: 2,4-dimethylhept-1-ene
Synonyms: 2,4-dimethylhept-1-ene
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: CZGAOHSMVSIJJZ-UHFFFAOYNA-N
SMILES: CCCC(C)CC(C)=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results