missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1,3-pentadiene, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 155360250

 View more versions of this product

Catalog No. AC155360250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 4-Dimethyl-1, 3-pentadiene
97.5
0.7400g/mL
10°C
98%
C7H12
(CH3)2C=CHC(CH3)=CH2
25g
0.74
CMSUNVGIWAFNBG-UHFFFAOYSA-N
2,4-dimethylpenta-1,3-diene
66080
98%
1000-86-8
100.0
94°C
Authentic
Glass bottle
1.4400 to 1.442
MFCD00008903
01, 257
2,4-dimethyl-1,3-pentadiene, 1,1,3-trimethylbutadiene, 1,3-pentadiene, 2,4-dimethyl, acmc-20aoci, 1,3-trimethylbutadiene, 1,3-pentadiene,2,4-dimethyl, cmsunvgiwafnbg-uhfffaoysa, ch3 2c=chc ch3 =ch2, 1,3-pentadiene, 2,4-dimethyl-8ci 9ci
CC(=CC(=C)C)C
96.17
96.17

Chemical Identifiers

1000-86-8
96.17
CMSUNVGIWAFNBG-UHFFFAOYSA-N
66080
CC(=CC(=C)C)C
C7H12
MFCD00008903
2,4-dimethyl-1,3-pentadiene, 1,1,3-trimethylbutadiene, 1,3-pentadiene, 2,4-dimethyl, acmc-20aoci, 1,3-trimethylbutadiene, 1,3-pentadiene,2,4-dimethyl, cmsunvgiwafnbg-uhfffaoysa, ch3 2c=chc ch3 =ch2, 1,3-pentadiene, 2,4-dimethyl-8ci 9ci
2,4-dimethylpenta-1,3-diene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-677-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only