missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1,3-pentadiene, 98%

Catalog No. p-3736334
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155360050 5 g Glass bottle
AC155360250 25 g Glass bottle
AC155361000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC155360050 Supplier Thermo Scientific Chemicals Supplier No. 155360050

CAS: 1000-86-8 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1000-86-8
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00008903
InChI Key CMSUNVGIWAFNBG-UHFFFAOYSA-N
Synonym 2,4-dimethyl-1,3-pentadiene, 1,1,3-trimethylbutadiene, 1,3-pentadiene, 2,4-dimethyl, acmc-20aoci, 1,3-trimethylbutadiene, 1,3-pentadiene,2,4-dimethyl, cmsunvgiwafnbg-uhfffaoysa, ch3 2c=chc ch3 =ch2, 1,3-pentadiene, 2,4-dimethyl-8ci 9ci
PubChem CID 66080
IUPAC Name 2,4-dimethylpenta-1,3-diene
SMILES CC(=CC(=C)C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7400g/mL
Boiling Point 94°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4400 to 1.442
Linear Formula (CH3)2C=CHC(CH3)=CH2
Quantity 5 g
Beilstein 01, 257
Specific Gravity 0.74
Formula Weight 96.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethyl-1, 3-pentadiene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-677-4

RUO – Research Use Only