missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1,3-dioxane 98.0+%, TCI America™

Catalog No. D070825ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D070825ML 25 mL
D070810ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D070825ML Supplier TCI America Supplier No. D070825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 766-20-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00023821
InChI Key VREPYGYMOSZTKJ-UHFFFAOYNA-N
Synonym 2,4-dimethyl-m-dioxane, 1,3-dioxane, 2,4-dimethyl, m-dioxane, 2,4-dimethyl, cis-2,4-dimethyl-1,3-dioxane, acmc-209p4w, 5-19-01-00072 beilstein handbook reference, 2,4-dimethyl-1,3 dioxane, 1,3-dioxane,2,4-dimethyl, trans-2,4-dimethyl-1,3-dioxane, m-dioxane, 2,4-dimethyl-8ci
PubChem CID 13009
IUPAC Name 2,4-dimethyl-1,3-dioxane
SMILES CC1CCOC(C)O1

Specifications

Color Colorless
Boiling Point 118°C
Quantity 25 mL
UN Number 2707
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethyl-1,3-dioxane
EINECSNumber (5)-0838
RTECSNumber JH0675000
TSCA No