accessibility menu, dialog, popup

UserName

CAS RN 766-20-1

IUPAC Name: 2,4-dimethyl-1,3-dioxane
Synonyms: 1,3-dioxane, 2,4-dimethyl-
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: VREPYGYMOSZTKJ-UHFFFAOYNA-N
SMILES: CC1CCOC(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results