missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyphenylacetic acid, 98%

Catalog No. AAL1378403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1378403 1 g
AAL1378406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1378403 Supplier Thermo Scientific Chemicals Supplier No. L1378403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6496-89-5 | C10H11O4 | 195.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6496-89-5
Molecular Formula C10H11O4
Molecular Weight (g/mol) 195.20
MDL Number MFCD00183328
InChI Key ZFXFMGARFHRTTO-UHFFFAOYSA-M
PubChem CID 350555
IUPAC Name 2-(2,4-dimethoxyphenyl)acetic acid
SMILES COC1=CC=C(CC([O-])=O)C(OC)=C1

Specifications

Melting Point 108°C to 112°C
Quantity 1 g
Beilstein 2694132
Synonym 2,4-dimethoxyphenylacetic acid, 2-2,4-dimethoxyphenyl acetic acid, 2,4-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,4-dimethoxy, 2,4-dimethoxybenzeneacetic acid, acmc-209nop, ksc497q2d, 2,4-dimethoxyphenylaceticacid, 2, 4-dimethoxyphenylacetic acid
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 2,4-Dimethoxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only