missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyiodobenzene 98.0+%, TCI America™

Catalog No. D063610G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D063610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D063610G Supplier TCI America Supplier No. D063610G

Chemical Identifiers

CAS 20469-63-0
Molecular Formula C8H9IO2
Molecular Weight (g/mol) 264.062
MDL Number MFCD00059268
InChI Key ZDUYJCKEORTAQE-UHFFFAOYSA-N
Synonym 2,4-dimethoxyiodobenzene, 2,4-dimethoxy-1-iodobenzene, 1,3-dimethoxy-4-iodobenzene, 1-iodo-2,4-dimethoxy-benzene, benzene, 1-iodo-2,4-dimethoxy, 4-iodo-1,3-dimethoxybenzene, pubchem3053, 4-iodo-3-methoxy anisole, dimethoxyiodobenzene, cambridge id 5107587
PubChem CID 140694
IUPAC Name 1-iodo-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)I)OC

Specifications

Melting Point 42°C
Color White
Boiling Point 166°C
Quantity 10 g
Formula Weight 264.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxyiodobenzene