missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzyl Alcohol 98.0+%, TCI America™

Catalog No. D18355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18355G 5 g
D183525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18355G Supplier TCI America Supplier No. D18355G

Chemical Identifiers

CAS 7314-44-5
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004614
InChI Key RNKOUSCCPHSCFE-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol
PubChem CID 81750
IUPAC Name (2,4-dimethoxyphenyl)methanol
SMILES COC1=CC(=C(C=C1)CO)OC

Specifications

Melting Point 36°C
Color White
Boiling Point 135°C
Quantity 5 g
Formula Weight 168.19
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,4-Dimethoxybenzyl Alcohol