missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzyl alcohol, 99%

Catalog No. AC170460250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170460250 25 g Glass bottle
AC170461000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170460250 Supplier Thermo Scientific Chemicals Supplier No. 170460250

CAS: 7314-44-5 | C9H12O3 | 168.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7314-44-5
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00004614
InChI Key RNKOUSCCPHSCFE-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol
PubChem CID 81750
IUPAC Name (2,4-dimethoxyphenyl)methanol
SMILES COC1=CC(=C(C=C1)CO)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 37.0°C to 40.0°C
Boiling Point 177.0°C to 179.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5450 to 1.5480
Linear Formula (CH3O)2C6H3CH2OH
Quantity 25 g
Beilstein 06, I, 550
Formula Weight 168.19
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 2, 4-Dimethoxybenzyl alcohol
Show More Show Less
EINECSNumber 230-775-2

RUO – Research Use Only