missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzonitrile 99.0+%, TCI America™

Catalog No. p-7128504
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48545G 5 g
D485425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48545G Supplier TCI America Supplier No. D48545G
Only null left

Chemical Identifiers

CAS 4107-65-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00001786
InChI Key RYRZSQQELLQCMZ-UHFFFAOYSA-N
Synonym benzonitrile, 2,4-dimethoxy, 2,4-dimethoxy-benzonitrile, 2,4-dimethoxybenzenecarbonitrile, pubchem13105, acmc-1anqc, 2.4-dimethoxybenzonitril, ksc497m9f, 2,4-dimethoxybenzonitrile
PubChem CID 77750
IUPAC Name 2,4-dimethoxybenzonitrile
SMILES COC1=CC(=C(C=C1)C#N)OC

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 163.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxybenzonitrile