missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzonitrile, 99%

CAS: 4107-65-7 | C9H9NO2 | 163.176 g/mol

$26.64 - $89.15

Chemical Identifiers

CAS 4107-65-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00001786
InChI Key RYRZSQQELLQCMZ-UHFFFAOYSA-N
Synonym benzonitrile, 2,4-dimethoxy, 2,4-dimethoxy-benzonitrile, 2,4-dimethoxybenzenecarbonitrile, pubchem13105, acmc-1anqc, 2.4-dimethoxybenzonitril, ksc497m9f, 2,4-dimethoxybenzonitrile
PubChem CID 77750
IUPAC Name 2,4-dimethoxybenzonitrile
SMILES COC1=CC(=C(C=C1)C#N)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2462106
View Documents
Thermo Scientific Chemicals
B2462106
5 g
Each for $26.64
Only null left
 
AAB2462114
View Documents
Thermo Scientific Chemicals
B2462114
25 g
Each for $89.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4107-65-7
163.176
RYRZSQQELLQCMZ-UHFFFAOYSA-N
77750
COC1=CC(=C(C=C1)C#N)OC
C9H9NO2
MFCD00001786
benzonitrile, 2,4-dimethoxy, 2,4-dimethoxy-benzonitrile, 2,4-dimethoxybenzenecarbonitrile, pubchem13105, acmc-1anqc, 2.4-dimethoxybenzonitril, ksc497m9f, 2,4-dimethoxybenzonitrile
2,4-dimethoxybenzonitrile

Specifications

4107-65-7
C9H9NO2
5 g
1950512
RYRZSQQELLQCMZ-UHFFFAOYSA-N
2,4-dimethoxybenzonitrile
77750
99%
92°C to 95°C
MFCD00001786
UN3439
benzonitrile, 2,4-dimethoxy, 2,4-dimethoxy-benzonitrile, 2,4-dimethoxybenzenecarbonitrile, pubchem13105, acmc-1anqc, 2.4-dimethoxybenzonitril, ksc497m9f, 2,4-dimethoxybenzonitrile
COC1=CC(=C(C=C1)C#N)OC
163.176
163.18
2,4-Dimethoxybenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 223-886-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only