missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzoic Acid 99.0+%, TCI America™

Catalog No. p-7126869
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16755G 5 g
D167525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16755G Supplier TCI America Supplier No. D16755G
Only null left

Chemical Identifiers

CAS 91-52-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00002434
InChI Key GPVDHNVGGIAOQT-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dimethoxy, 2,4-dimethoxybenzoicacid, 2,4-dimethoxy benzoic acid, 2,4-dimethoxybenzoic, pubchem4949, rarechem al bo 0028, 2,4-dimethoxy-benzoicaci, 2,4 dimethoxybenzoic acid, 2.4-dimethoxybenzoic acid, dimethoxybenzoic-2,4 acid
PubChem CID 7052
IUPAC Name 2,4-dimethoxybenzoic acid
SMILES COC1=CC(=C(C=C1)C(=O)O)OC

Specifications

Melting Point 110°C
Color White
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxybenzoic Acid