Learn More
2,4-Dimethoxybenzoic acid, 98%
CAS: 91-52-1 | C9H10O4 | 182.175 g/mol
Supplier: Thermo Scientific Chemicals A1625214
Description
Yttrium and heavy lanthanide complexes is synthesised with 2, 4-dimethoxybenzoic acid and thermal and spectral features is studied. In the case of the electron-rich aromatic system 2,4-dimethoxybenzoic acid reduction of the intermediate HOBt ester is slow, and good.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsYttrium and heavy lanthanide complexes is synthesised with 2, 4-dimethoxybenzoic acid and thermal and spectral features is studied. In the case of the electron-rich aromatic system 2,4-dimethoxybenzoic acid reduction of the intermediate HOBt ester is slow, and good.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| 2,4-Dimethoxybenzoic acid | |
| 107°C to 111°C | |
| C9H10O4 | |
| 25 g | |
| benzoic acid, 2,4-dimethoxy, 2,4-dimethoxybenzoicacid, 2,4-dimethoxy benzoic acid, 2,4-dimethoxybenzoic, pubchem4949, rarechem al bo 0028, 2,4-dimethoxy-benzoicaci, 2,4 dimethoxybenzoic acid, 2.4-dimethoxybenzoic acid, dimethoxybenzoic-2,4 acid | |
| GPVDHNVGGIAOQT-UHFFFAOYSA-N | |
| 2,4-dimethoxybenzoic acid | |
| 7052 | |
| 98% |
| 91-52-1 | |
| Odorless | |
| MFCD00002434 | |
| 2210860 | |
| Slightly soluble in water. | |
| COC1=CC(=C(C=C1)C(=O)O)OC | |
| 182.175 | |
| 182.18 |
Chemical Identifiers
| 91-52-1 | |
| 182.175 | |
| GPVDHNVGGIAOQT-UHFFFAOYSA-N | |
| 7052 | |
| COC1=CC(=C(C=C1)C(=O)O)OC |
| C9H10O4 | |
| MFCD00002434 | |
| benzoic acid, 2,4-dimethoxy, 2,4-dimethoxybenzoicacid, 2,4-dimethoxy benzoic acid, 2,4-dimethoxybenzoic, pubchem4949, rarechem al bo 0028, 2,4-dimethoxy-benzoicaci, 2,4 dimethoxybenzoic acid, 2.4-dimethoxybenzoic acid, dimethoxybenzoic-2,4 acid | |
| 2,4-dimethoxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-074-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only