missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethoxyacetophenone 96.0+%, TCI America™

Catalog No. D203025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D203025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D203025G Supplier TCI America Supplier No. D203025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 829-20-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00008727
InChI Key VQTDPCRSXHFMOL-UHFFFAOYSA-N
Synonym 2',4'-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethanone, 2,4-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 1-2,4-dimethoxyphenyl, acetophenone, 2',4'-dimethoxy, peonol methyl ether, 1-acetyl-2,4-dimethoxybenzene, pubchem10582, acmc-209pqs
PubChem CID 70016
IUPAC Name 1-(2,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)OC

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 297°C
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,4′-Dimethoxyacetophenone
EINECSNumber (3)-1253
TSCA No