accessibility menu, dialog, popup

UserName

H3COCH3OOH3C

CAS RN 829-20-9

IUPAC Name: 1-(2,4-dimethoxyphenyl)ethan-1-one
Synonyms: 1-(2,4-dimethoxyphenyl)ethanone
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: VQTDPCRSXHFMOL-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(C)=O)C(OC)=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)